accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 22409-91-2

IUPAC Name: ethyl 3-phenoxypropanoate
Synonyms: ethyl 3-phenoxypropanoate
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: TXKJOPXGYSFUNC-UHFFFAOYSA-N
SMILES: CCOC(=O)CCOC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results