accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOOHOOHOHONH3NH3NH3

CAS RN 22411-24-1

IUPAC Name: 4-sulfobenzene-1,2-dicarboxylic acid triamine
Synonyms: 4-sulfobenzene-1,2-dicarboxylic acid triamine
Molecular Weight (g/mol): 297.28
Molecular Formula: C8H15N3O7S
InChi Key: RMPQIFXEQFYSEM-UHFFFAOYSA-N
SMILES: N.N.N.OC(=O)C1=C(C=C(C=C1)S(O)(=O)=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results