accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBrBrLa

CAS RN 224183-16-8

IUPAC Name: lanthanum(3+) tribromide
Synonyms: lanthanum(III) bromide
Molecular Weight (g/mol): 378.62
Molecular Formula: Br3La
InChi Key: XKUYOJZZLGFZTC-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[La+3]
Grade 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)