accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22426-14-8

IUPAC Name: 2-bromo-1,10-phenanthroline
Synonyms: 2-bromo-1,10-phenanthroline
Molecular Weight (g/mol): 259.11
Molecular Formula: C12H7BrN2
InChi Key: ZRJUDAZGVGIDLP-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=CC3=CC=CN=C3C2=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)