accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CN

CAS RN 2243-27-8

IUPAC Name: nonanenitrile
Synonyms: nonanonitrile
Molecular Weight (g/mol): 139.24
Molecular Formula: C9H17N
InChi Key: PLZZPPHAMDJOSR-UHFFFAOYSA-N
SMILES: CCCCCCCCC#N
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)