accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH2NNH

CAS RN 2243-56-3

IUPAC Name: hydrogen (naphthalen-1-yl)hydrazine chloride
Synonyms: hydrogen 1-naphthylhydrazine chloride
Molecular Weight (g/mol): 194.66
Molecular Formula: C10H11ClN2
InChi Key: FYSSYOCJFZSKNW-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results