accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2243-62-1

IUPAC Name: naphthalene-1,5-diamine
Synonyms: 1,5-diaminonaphthalene 1,5-naphthylenediamine
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: KQSABULTKYLFEV-UHFFFAOYSA-N
SMILES: NC1=CC=CC2=C(N)C=CC=C12
Percent Purity 
  • (3)
  • (1)
  • (4)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)