accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2243-76-7

IUPAC Name: 3-[2-(4-nitrophenyl)hydrazin-1-ylidene]-6-oxocyclohexa-1,4-diene-1-carboxylic acid
Synonyms: 3-[2-(4-nitrophenyl)hydrazin-1-ylidene]-6-oxocyclohexa-1,4-diene-1-carboxylic acid
Molecular Weight (g/mol): 287.23
Molecular Formula: C13H9N3O5
InChi Key: RTQYUQUDISIJCV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(C=CC1=O)=NNC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results