accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH

CAS RN 2243-98-3

IUPAC Name: undec-1-yne
Synonyms: 1-undecyne
Molecular Weight (g/mol): 152.28
Molecular Formula: C11H20
InChi Key: YVSFLVNWJIEJRV-UHFFFAOYSA-N
SMILES: CCCCCCCCCC#C
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)