accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONHFFF

CAS RN 2249-28-7

IUPAC Name: 4-[3-(trifluoromethyl)phenyl]piperidin-4-ol
Synonyms: 4-[3-(trifluoromethyl)phenyl]piperidin-4-ol
Molecular Weight (g/mol): 245.25
Molecular Formula: C12H14F3NO
InChi Key: ILKPZCKFWLTEBQ-UHFFFAOYSA-N
SMILES: OC1(CCNCC1)C1=CC=CC(=C1)C(F)(F)F
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)