accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONOO

CAS RN 22509-74-6

IUPAC Name: ethyl 1,3-dioxo-2,3-dihydro-1H-isoindole-2-carboxylate
Synonyms: N-carbethoxyphthalimide
Molecular Weight (g/mol): 219.2
Molecular Formula: C11H9NO4
InChi Key: VRHAQNTWKSVEEC-UHFFFAOYSA-N
SMILES: CCOC(=O)N1C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (1)
  • (5)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)