accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFOBr

CAS RN 2252-44-0

IUPAC Name: 1-bromo-3-(trifluoromethoxy)benzene
Synonyms: 1-bromo-3-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 241.01
Molecular Formula: C7H4BrF3O
InChi Key: WVUDHWBCPSXAFN-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC(Br)=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)