accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22535-90-6

IUPAC Name: 6-(2-{9-[2-(4,6-diamino-1,3,5-triazin-2-yl)ethyl]-2,4,8,10-tetraoxaspiro[5.5]undecan-3-yl}ethyl)-1,3,5-triazine-2,4-diamine
Synonyms: 6-(2-{9-[2-(4,6-diamino-1,3,5-triazin-2-yl)ethyl]-2,4,8,10-tetraoxaspiro[5.5]undecan-3-yl}ethyl)-1,3,5-triazine-2,4-diamine
Molecular Weight (g/mol): 434.46
Molecular Formula: C17H26N10O4
InChi Key: DUZLHGMYNVZMCO-UHFFFAOYSA-N
SMILES: NC1=NC(CCC2OCC3(CO2)COC(CCC2=NC(N)=NC(N)=N2)OC3)=NC(N)=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results