accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNCl

CAS RN 22536-61-4

IUPAC Name: 2-chloro-5-methylpyrimidine
Synonyms: 2-chloro-5-methylpyrimidine
Molecular Weight (g/mol): 128.56
Molecular Formula: C5H5ClN2
InChi Key: APRMCBSTMFKLEI-UHFFFAOYSA-N
SMILES: CC1=CN=C(Cl)N=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)