CAS RN 2254-88-8
IUPAC Name:
dichloro-1,2,4-thiadiazole
Synonyms:
dichloro-1,2,4-thiadiazole
Molecular Weight (g/mol):
155
Molecular Formula:
C2Cl2N2S
InChi Key:
CEUHPOVLEQUFCC-UHFFFAOYSA-N
SMILES:
ClC1=NC(Cl)=NS1
1
–
1
of
1
results
1
Promotions Available
3,5-Dichloro-1,2,4-thiadiazole, 97%, Thermo Scientific Chemicals
3,5-Dichloro-1,2,4-thiadiazole, 97%, Thermo Scientific Chemicals
CAS: 2254-88-8 Molecular Formula: C2Cl2N2S Molecular Weight (g/mol): 155.01 InChI Key: CEUHPOVLEQUFCC-UHFFFAOYSA-N PubChem CID: 2725055 IUPAC Name: 3,5-dichloro-1,2,4-thiadiazole SMILES: C1(=NSC(=N1)Cl)Cl