accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNSS

CAS RN 2254-94-6

IUPAC Name: 3-methyl-2,3-dihydro-1,3-benzothiazole-2-thione
Synonyms: 3-methyl-1,3-benzothiazole-2-thione
Molecular Weight (g/mol): 181.27
Molecular Formula: C8H7NS2
InChi Key: IRNRNPNZAKHEAW-UHFFFAOYSA-N
SMILES: CN1C(=S)SC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results