accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22551-24-2

IUPAC Name: 2-(methylsulfanyl)acetamide
Synonyms: 2-(methylsulfanyl)acetamide
Molecular Weight (g/mol): 105.16
Molecular Formula: C3H7NOS
InChi Key: OBENGAMZEGRSIC-UHFFFAOYSA-N
SMILES: CSCC(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results