accessibility menu, dialog, popup

UserName

Viewing profile as user:
BCH3H3CH3CK

CAS RN 22560-21-0

IUPAC Name: potassium triethyl-λ²-boranuide
Synonyms: potassium triethyl-λ²-boranuide
Molecular Weight (g/mol): 137.09
Molecular Formula: C6H15BK
InChi Key: JSWZHFBULBZRJI-UHFFFAOYSA-N
SMILES: [K+].CC[B-](CC)CC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results