accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22589-35-1

IUPAC Name: 4-(thiomorpholin-4-yl)aniline
Synonyms: 4-(thiomorpholin-4-yl)aniline
Molecular Weight (g/mol): 194.3
Molecular Formula: C10H14N2S
InChi Key: RQXQMUUSQGCLPL-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)N1CCSCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results