accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2260-50-6

IUPAC Name: [2-(acetyloxy)ethyl]trimethylazanium iodide
Synonyms: acetylcholine iodide
Molecular Weight (g/mol): 273.11
Molecular Formula: C7H16INO2
InChi Key: SMBBQHHYSLHDHF-UHFFFAOYSA-M
SMILES: [I-].CC(=O)OCC[N+](C)(C)C
Percent Purity 
  • (1)
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (4)