CAS RN 22612-89-1
IUPAC Name:
1,1,3,3-tetrabromopropan-2-one
Synonyms:
1,1,3,3-tetrabromopropan-2-one
Molecular Weight (g/mol):
373.66
Molecular Formula:
C3H2Br4O
InChi Key:
SAMNBOHOBWEEEU-UHFFFAOYSA-N
SMILES:
BrC(Br)C(=O)C(Br)Br
1
–
1
of
1
results
1
1,1,3,3-Tetrabromoacetone 98.0+%, TCI America™
CAS: 22612-89-1 Molecular Formula: C3H2Br4O Molecular Weight (g/mol): 373.664 MDL Number: MFCD00059470 InChI Key: SAMNBOHOBWEEEU-UHFFFAOYSA-N Synonym: 1,1,3,3-Tetrabromo-2-propanone PubChem CID: 608441 IUPAC Name: 1,1,3,3-tetrabromopropan-2-one SMILES: C(C(=O)C(Br)Br)(Br)Br