accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 226396-31-2

IUPAC Name: [4-(methylsulfamoyl)phenyl]boronic acid
Synonyms: 4-(methylsulfamoyl)phenylboronic acid
Molecular Weight (g/mol): 215.03
Molecular Formula: C7H10BNO4S
InChi Key: DOQOQZHSIBBHMY-UHFFFAOYSA-N
SMILES: CNS(=O)(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results