accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2265-93-2

IUPAC Name: 2,4-difluoro-1-iodobenzene
Synonyms: 2,4-difluoro-1-iodobenzene
Molecular Weight (g/mol): 239.99
Molecular Formula: C6H3F2I
InChi Key: YKLDMAPEGQYZRT-UHFFFAOYSA-N
SMILES: FC1=CC=C(I)C(F)=C1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)