accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22661-87-6

IUPAC Name: hydrogen N''-methylguanidine chloride
Synonyms: hydrogen methyl-guanidine chloride
Molecular Weight (g/mol): 109.56
Molecular Formula: C2H8ClN3
InChi Key: VJQCNCOGZPSOQZ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN=C(N)N
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)