accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONNNBr

CAS RN 226929-39-1

IUPAC Name: 4-[1-({1-[(4-bromophenyl)methyl]-1H-imidazol-5-yl}methyl)-4-(naphthalen-1-yl)-1H-pyrrole-3-carbonyl]morpholine
Synonyms: 4-[1-({3-[(4-bromophenyl)methyl]imidazol-4-yl}methyl)-4-(naphthalen-1-yl)pyrrole-3-carbonyl]morpholine
Molecular Weight (g/mol): 555.48
Molecular Formula: C30H27BrN4O2
InChi Key: GUUIRIMAQGOLHT-UHFFFAOYSA-N
SMILES: BrC1=CC=C(CN2C=NC=C2CN2C=C(C(=O)N3CCOCC3)C(=C2)C2=C3C=CC=CC3=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results