accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNCH3

CAS RN 226958-06-1

IUPAC Name: 9-[7-(9H-carbazol-9-yl)-9,9-dimethyl-9H-fluoren-2-yl]-9H-carbazole
Synonyms: 9-[7-(carbazol-9-yl)-9,9-dimethylfluoren-2-yl]carbazole
Molecular Weight (g/mol): 524.67
Molecular Formula: C39H28N2
InChi Key: IEQGNDONCZPWMW-UHFFFAOYSA-N
SMILES: CC1(C)C2=C(C=CC(=C2)N2C3=C(C=CC=C3)C3=C2C=CC=C3)C2=C1C=C(C=C2)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)