accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22717-57-3

IUPAC Name: methyl 2-hydroxy-5-methylbenzoate
Synonyms: methyl 2-hydroxy-5-methylbenzoate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: JQYUQKRFSSSGJM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(C)=CC=C1O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results