accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22718-49-6

IUPAC Name: (dimethylamino)urea
Synonyms: 4,4-dimethylsemicarbazide
Molecular Weight (g/mol): 103.13
Molecular Formula: C3H9N3O
InChi Key: SVZFWZRCMLRFOM-UHFFFAOYSA-N
SMILES: CN(C)NC(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results