accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaAlH3COOH3COO

CAS RN 22722-98-1

IUPAC Name: alumanylium sodium bis(2-methoxyethan-1-olate)
Synonyms: alumanylium sodium bis(2-methoxyethanolate)
Molecular Weight (g/mol): 202.16
Molecular Formula: C6H16AlNaO4
InChi Key: XJIQVZMZXHEYOY-UHFFFAOYSA-N
SMILES: [Na+].[AlH2+].COCC[O-].COCC[O-]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (5)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (2)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)