accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3COH2N

CAS RN 22737-36-6

IUPAC Name: O-(trimethylsilyl)hydroxylamine
Synonyms: O-(trimethylsilyl)hydroxylamine
Molecular Weight (g/mol): 105.21
Molecular Formula: C3H11NOSi
InChi Key: AEKHNNJSMVVESS-UHFFFAOYSA-N
SMILES: C[Si](C)(C)ON
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)