accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClClHNOOHClClCl

CAS RN 2277-92-1

IUPAC Name: 2,3,5-trichloro-N-(3,5-dichloro-2-hydroxyphenyl)-6-hydroxybenzamide
Synonyms: oxyclozanide
Molecular Weight (g/mol): 401.45
Molecular Formula: C13H6Cl5NO3
InChi Key: JYWIYHUXVMAGLG-UHFFFAOYSA-N
SMILES: OC1=C(C(=O)NC2=CC(Cl)=CC(Cl)=C2O)C(Cl)=C(Cl)C=C1Cl
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)