accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2N

CAS RN 22795-99-9

IUPAC Name: 1-(1-ethylpyrrolidin-2-yl)methanamine
Synonyms: 1-(1-ethylpyrrolidin-2-yl)methanamine
Molecular Weight (g/mol): 128.22
Molecular Formula: C7H16N2
InChi Key: UNRBEYYLYRXYCG-UHFFFAOYNA-N
SMILES: CCN1CCCC1CN
Molecular Weight (g/mol) 
  • (4)
  • (2)
  • (2)
Quantity 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)