accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3COH(S)NH2OHO

CAS RN 2280-27-5

IUPAC Name: (2S)-2-amino-3-hydroxy-3-methylbutanoic acid
Synonyms: (2S)-2-amino-3-hydroxy-3-methylbutanoic acid
Molecular Weight (g/mol): 133.15
Molecular Formula: C5H11NO3
InChi Key: LDRFQSZFVGJGGP-GSVOUGTGSA-N
SMILES: CC(C)(O)[C@H](N)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)