accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22809-37-6

IUPAC Name: 6-bromohexanoyl chloride
Synonyms: 6-bromohexanoyl chloride
Molecular Weight (g/mol): 213.5
Molecular Formula: C6H10BrClO
InChi Key: HBPVGJGBRWIVSX-UHFFFAOYSA-N
SMILES: ClC(=O)CCCCCBr
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)