accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CH3CPCH3PH3CCH3ClClRu(?)

CAS RN 228120-95-4

IUPAC Name: λ²-ruthenium(2+) 1-methyl-4-(propan-2-yl)benzene {2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane dichloride
Synonyms: λ²-ruthenium(2+) cymene {2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane dichloride
Molecular Weight (g/mol): 984.99
Molecular Formula: C58H54Cl2P2Ru
InChi Key: BHIUOOHVIWKUTN-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Ru++].CC(C)C1=CC=C(C)C=C1.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results