accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22817-26-1

IUPAC Name: 3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene
Synonyms: 3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene
Molecular Weight (g/mol): 169.23
Molecular Formula: C12H11N
InChi Key: NBWCGDRKSOXZNI-UHFFFAOYSA-N
SMILES: C1NCC2=C3C1=CC=CC3=CC=C2
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results