accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHO

CAS RN 2283-08-1

IUPAC Name: 2-hydroxynaphthalene-1-carboxylic acid
Synonyms: β-hydroxynaphthoic acid
Molecular Weight (g/mol): 188.18
Molecular Formula: C11H8O3
InChi Key: UPHOPMSGKZNELG-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC=CC2=CC=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results