accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CPNH3CNCH3CH3H3C

CAS RN 2283-11-6

IUPAC Name: [bis(diethylamino)phosphanyl]diethylamine
Synonyms: [bis(diethylamino)phosphanyl]diethylamine
Molecular Weight (g/mol): 247.37
Molecular Formula: C12H30N3P
InChi Key: FDIOSTIIZGWENY-UHFFFAOYSA-N
SMILES: CCN(CC)P(N(CC)CC)N(CC)CC
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (7)
Quantity 
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (7)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (5)