CAS RN 22839-47-0
Thermo Scientific Chemicals Aspartame, 98%
CAS: 22839-47-0 Molecular Formula: C14H18N2O5 Molecular Weight (g/mol): 294.31 MDL Number: MFCD00002724 InChI Key: IAOZJIPTCAWIRG-QWRGUYRKSA-N Synonym: aspartame,nutrasweet,asp-phe-ome,asp-phe methyl ester,aspartam,aspartamum,aspartamo,l-aspartyl-l-phenylalanine methyl ester,aspartylphenylalanine methyl ester,dipeptide sweetener PubChem CID: 134601 ChEBI: CHEBI:2877 IUPAC Name: (3S)-3-amino-4-[[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]amino]-4-oxobutanoic acid SMILES: COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O
Thermo Scientific Chemicals L-Aspartyl-L-phenylalanine methyl ester, 98%
CAS: 22839-47-0 Molecular Formula: C14H18N2O5 Molecular Weight (g/mol): 294.31 MDL Number: MFCD00002724 InChI Key: IAOZJIPTCAWIRG-QWRGUYRKSA-N Synonym: aspartame,nutrasweet,asp-phe-ome,asp-phe methyl ester,aspartam,aspartamum,aspartamo,l-aspartyl-l-phenylalanine methyl ester,aspartylphenylalanine methyl ester,dipeptide sweetener PubChem CID: 134601 ChEBI: CHEBI:2877 IUPAC Name: (3S)-3-amino-4-[[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]amino]-4-oxobutanoic acid SMILES: COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O
Aspartame 98.0+%, TCI America™
CAS: 22839-47-0 Molecular Formula: C14H18N2O5 Molecular Weight (g/mol): 294.31 MDL Number: MFCD00002724 InChI Key: IAOZJIPTCAWIRG-QWRGUYRKSA-N Synonym: aspartame,nutrasweet,asp-phe-ome,asp-phe methyl ester,aspartam,aspartamum,aspartamo,l-aspartyl-l-phenylalanine methyl ester,aspartylphenylalanine methyl ester,dipeptide sweetener PubChem CID: 134601 ChEBI: CHEBI:2877 IUPAC Name: (3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid SMILES: COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O
Aspartame, Powder, NF, 98-102%, Spectrum™ Chemical
CAS: 22839-47-0 Molecular Formula: C14H18N2O5 Molecular Weight (g/mol): 294.31 MDL Number: MFCD00002724 InChI Key: IAOZJIPTCAWIRG-QWRGUYRKSA-N IUPAC Name: (3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid SMILES: COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O
Aspartame, Powder, NF, 2.5 kg, Spectrum Chemical
CAS: 22839-47-0 Molecular Formula: C14H18N2O5 Molecular Weight (g/mol): 294.31 MDL Number: MFCD00002724 InChI Key: IAOZJIPTCAWIRG-QWRGUYRKSA-N IUPAC Name: (3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid SMILES: COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O
Aspartame, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Aspartame is a low-calorie nutritive sugar substitute used in table-top sweeteners, chewing gums, soft drinks, etc.
L-Aspartyl-L-Phenylalanine methyl ester, 99.8%, MP Biomedicals™
CAS: 22839-47-0 Molecular Formula: C14H18N2O5 Molecular Weight (g/mol): 294.31 MDL Number: MFCD00002724 InChI Key: IAOZJIPTCAWIRG-QWRGUYRKSA-N Synonym: aspartame,nutrasweet,asp-phe-ome,asp-phe methyl ester,aspartam,aspartamum,aspartamo,l-aspartyl-l-phenylalanine methyl ester,aspartylphenylalanine methyl ester,dipeptide sweetener PubChem CID: 134601 ChEBI: CHEBI:2877 IUPAC Name: (3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid SMILES: COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O
Aspartame, Powder, FCC, 98-102%, Spectrum™ Chemical
CAS: 22839-47-0 Molecular Formula: C14H18N2O5 Molecular Weight (g/mol): 294.31 MDL Number: MFCD00002724 InChI Key: IAOZJIPTCAWIRG-QWRGUYRKSA-N IUPAC Name: (3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid SMILES: COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O