accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22863-82-7

IUPAC Name: (anthracen-2-yl)methanol
Synonyms: anthracen-2-ylmethanol
Molecular Weight (g/mol): 208.26
Molecular Formula: C15H12O
InChi Key: ZMUUBYLNJMTHBS-UHFFFAOYSA-N
SMILES: OCC1=CC2=CC3=CC=CC=C3C=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results