accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHN

CAS RN 22867-74-9

IUPAC Name: 7-ethyl-1H-indole
Synonyms: 7-ethyl-1H-indole
Molecular Weight (g/mol): 145.21
Molecular Formula: C10H11N
InChi Key: PIIZLMYXLGYWTN-UHFFFAOYSA-N
SMILES: CCC1=C2NC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results