accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22876-20-6

IUPAC Name: 6-chloro-2,3-dihydro-1,3-benzoxazole-2-thione
Synonyms: 6-chloro-3H-1,3-benzoxazole-2-thione
Molecular Weight (g/mol): 185.63
Molecular Formula: C7H4ClNOS
InChi Key: HAASPZUBSZGCKU-UHFFFAOYSA-N
SMILES: ClC1=CC=C2NC(=S)OC2=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)