accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22884-10-2

IUPAC Name: 2-(1H-imidazol-1-yl)acetic acid
Synonyms: imidazoleacetic acid
Molecular Weight (g/mol): 126.12
Molecular Formula: C5H6N2O2
InChi Key: QAFBDRSXXHEXGB-UHFFFAOYSA-N
SMILES: OC(=O)CN1C=CN=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results