accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 229-87-8

IUPAC Name: phenanthridine
Synonyms: phenanthridine 3,4-benzoquinoline 3,4-benzoisoquinoline 6-phenanthridine benzo(c)quinoline
Molecular Weight (g/mol): 179.22
Molecular Formula: C13H9N
InChi Key: RDOWQLZANAYVLL-UHFFFAOYSA-N
SMILES: C1=CC=C2C(=C1)C=NC1=CC=CC=C21
Molecular Weight (g/mol) 
  • (4)
  • (2)
Boiling Point 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)