accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22900-83-0

IUPAC Name: ethyl 2-bromo-4-methyl-1,3-thiazole-5-carboxylate
Synonyms: ethyl 2-bromo-4-methyl-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 250.11
Molecular Formula: C7H8BrNO2S
InChi Key: CFBIOWPDDZPIDP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N=C(Br)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results