CAS RN 22910-60-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Ginkgolic Acid (15:1)
A cell-permeable anacardic acid analog that inhibits protein SUMO .