CAS RN 22913-24-2
IUPAC Name:
methyl 1-benzothiophene-2-carboxylate
Synonyms:
methyl 1-benzothiophene-2-carboxylate
Molecular Weight (g/mol):
192.23
Molecular Formula:
C10H8O2S
InChi Key:
KRRAZMUPVIGDCU-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC2=CC=CC=C2S1
1
–
1
of
1
results
1
Sigma Aldrich Methyl benzo[b]thiophene-2-carboxylate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.