accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNBr

CAS RN 22918-01-0

IUPAC Name: 2-bromo-4-chloropyridine
Synonyms: 2-bromo-4-chloropyridine
Molecular Weight (g/mol): 192.44
Molecular Formula: C5H3BrClN
InChi Key: SURKZMFXICWLHU-UHFFFAOYSA-N
SMILES: ClC1=CC=NC(Br)=C1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)