accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 229180-65-8

POOHOOOHHOONH(R)

CAS RN 229180-65-8

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[4-(phosphonomethyl)phenyl]propanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-[4-(phosphonomethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 481.44
Molecular Formula: C25H24NO7P
InChi Key: OBRYXGOVBZKHLL-HSZRJFAPSA-N
SMILES: OC(=O)[C@@H](CC1=CC=C(CP(O)(O)=O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)