accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22948-06-7

IUPAC Name: 4-(triphenylmethyl)aniline
Synonyms: 4-(triphenylmethyl)aniline
Molecular Weight (g/mol): 335.45
Molecular Formula: C25H21N
InChi Key: XYHDHXBLSLSXSR-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)